4-[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride ≥97%
Supplier: Matrix Scientific
115119-500MG
115119-1G
102898-980EA
693.85
USD
102898-980
102898-982
4-[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride ≥97%
4-[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₈ClN₃O |
MDL Number:
MFCD28023521 CAS Number: 263383-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...