1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride ≥97%

Supplier: Matrix Scientific

115118-500MG 115118-1G
102898-976EA 693.85 USD
102898-976 102898-978
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride ≥97%
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂N₅O
MDL Number: MFCD28023520
CAS Number: 1239748-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR