1-{[3-(4-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine ≥97%

Supplier: Matrix Scientific

115110-500MG 115110-1G
102895-432EA 693.85 USD
102895-432 102895-434
1-{[3-(4-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine ≥97%
1-{[3-(4-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine

Hazard: Irritant

Formula: C₁₃H₁₅FN₄O
MDL Number: MFCD12655045
CAS Number: 1098360-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR