3-(6,8-Dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

115109-500MG 115109-1G
102895-428EA 693.85 USD
102895-428 102895-430
3-(6,8-Dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)propanoic acid ≥95%
3-(6,8-Dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₆
MW: 278.22 g/mol
MDL Number: MFCD14726785
CAS Number: 892293-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR