1-Pentyl-1H-indole-3-carboxylic acid 90

Supplier: Matrix Scientific

115107-1G 115107-500MG
102895-422EA 1040 USD
102895-422 102898-972
1-Pentyl-1H-indole-3-carboxylic acid 90
1-Pentyl-1H-indole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₂
MW: 231.3 g/mol
MDL Number: MFCD05859192
CAS Number: 727421-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR