4-Chloro-2-(3,4-dimethoxyphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde ≥90%

Supplier: Matrix Scientific

115106-500MG 115106-1G
102898-968EA 693.85 USD
102898-968 102898-970
4-Chloro-2-(3,4-dimethoxyphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde ≥90%
4-Chloro-2-(3,4-dimethoxyphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₁₂ClN₃O₃
MDL Number: MFCD22549263
CAS Number: 1544841-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR