Dimethyl-1-(2-oxo-2-phenylethyl)-1H-1,2,3-triazole-4,5-dicarboxylate ≥97%

Supplier: Matrix Scientific

115103-500MG 115103-1G
102895-416EA 693.85 USD
102895-416 102895-418
Dimethyl-1-(2-oxo-2-phenylethyl)-1H-1,2,3-triazole-4,5-dicarboxylate ≥97%
Dimethyl-1-(2-oxo-2-phenylethyl)-1H-1,2,3-triazole-4,5-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₁₃N₃O₅
MDL Number: MFCD22549280
CAS Number: 869895-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR