1'H-Spiro[cyclohexane-1,2'-quinazoline]-4'-thiol ≥97%
Supplier: Matrix Scientific
115102-500MG
115102-1G
102895-412EA
693.85
USD
102895-412
102895-414
1'H-Spiro[cyclohexane-1,2'-quinazoline]-4'-thiol ≥97%
1'H-Spiro[cyclohexane-1,2'-quinazoline]-4'-thiol
Hazard: Irritant
Formula:
C₁₃H₁₆N₂S |
MDL Number:
MFCD28023515 CAS Number: 126492-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...