1'H-Spiro[cycloheptane-1,2'-quinazoline]-4'-thiol ≥97%

Supplier: Matrix Scientific

115099-500MG 115099-1G
102891-610EA 693.85 USD
102891-610 102895-402
1'H-Spiro[cycloheptane-1,2'-quinazoline]-4'-thiol ≥97%
1'H-Spiro[cycloheptane-1,2'-quinazoline]-4'-thiol

Hazard: Irritant

Formula: C₁₄H₁₈N₂S
MDL Number: MFCD28023513
CAS Number: 892299-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR