2-{[(4-Methylphenyl)amino]methyl}phenol ≥97%

Supplier: Matrix Scientific

115098-500MG 115098-1G
102891-606EA 693.85 USD
102891-606 102891-608
2-{[(4-Methylphenyl)amino]methyl}phenol ≥97%
2-{[(4-Methylphenyl)amino]methyl}phenol

Hazard: Irritant

Formula: C₁₄H₁₅NO
MDL Number: MFCD00086756
CAS Number: 14674-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR