3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one ≥97%

Supplier: Matrix Scientific

115097-500MG 115097-1G
102891-602EA 693.85 USD
102891-602 102891-604
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one ≥97%
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one

Hazard: Irritant

Formula: C₁₈H₁₄N₂O₃S
MDL Number: MFCD07655393
CAS Number: 892300-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR