Dimethyl-1-(2-oxopropyl)1H-1,2,3-triazole-4,5-dicarboxylate ≥97%

Supplier: Matrix Scientific

115096-500MG 115096-1G
102891-598EA 693.85 USD
102891-598 102891-600
Dimethyl-1-(2-oxopropyl)1H-1,2,3-triazole-4,5-dicarboxylate ≥97%
Dimethyl-1-(2-oxopropyl)1H-1,2,3-triazole-4,5-dicarboxylate

Hazard: Irritant

Formula: C₉H₁₁N₃O₅
MW: 241.21 g/mol
MDL Number: MFCD22549283
CAS Number: 1039766-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR