1-(3-Aminophenyl)-5-oxo-4,5-dihydro-1H-pyrazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

115095-500MG 115095-1G
102891-594EA 693.85 USD
102891-594 102891-596
1-(3-Aminophenyl)-5-oxo-4,5-dihydro-1H-pyrazole-3-carboxylic acid ≥97%
1-(3-Aminophenyl)-5-oxo-4,5-dihydro-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉N₃O₃
MDL Number: MFCD00043812
CAS Number: 89-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR