1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]-1H-imidazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

115094-500MG 115094-1G
102891-590EA 693.85 USD
102891-590 102891-592
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]-1H-imidazole-4-carboxylic acid ≥97%
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]-1H-imidazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₉N₅O₃
MDL Number: MFCD16629529
CAS Number: 1239851-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR