3-{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}quinazoline-2,4(1H,3H)-dione ≥95%

Supplier: Matrix Scientific

115081-500MG 115081-1G
102895-370EA 693.85 USD
102895-370 102895-372
3-{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}quinazoline-2,4(1H,3H)-dione ≥95%
3-{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}quinazoline-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₈H₁₄N₄O₃
MDL Number: MFCD14703552
CAS Number: 1223889-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR