4-(4-Fluorophenyl)-2-(3-methoxyphenyl)-1H-imidazole-5-thiol ≥90%

Supplier: Matrix Scientific

115080-500MG 115080-1G
102895-366EA 693.85 USD
102895-366 102895-368
4-(4-Fluorophenyl)-2-(3-methoxyphenyl)-1H-imidazole-5-thiol ≥90%
4-(4-Fluorophenyl)-2-(3-methoxyphenyl)-1H-imidazole-5-thiol

Hazard: Irritant

Formula: C₁₆H₁₃FN₂OS
MDL Number: MFCD22549298
CAS Number: 1326832-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR