5-Amino-4H-1,2,4-triazole-3-carboxylic acid ≥90%

Supplier: Matrix Scientific

Synonyms: 3-Amino-1,2,4-triazole-5-carboxylic acid Hemihydrate

115079-500MG 115079-1G
102895-362EA 693.85 USD
102895-362 102895-364
5-Amino-4H-1,2,4-triazole-3-carboxylic acid ≥90%
5-Amino-4H-1,2,4-triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₃H₄N₄O₂
MW: 128.09 g/mol
MDL Number: MFCD00010620
CAS Number: 3641-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR