3-Phenyl-1,4,8-triazaspiro[4.5]dec-3-en-2-one ≥95%

Supplier: Matrix Scientific

115077-500MG 115077-1G
102898-954EA 693.85 USD
102898-954 102898-956
3-Phenyl-1,4,8-triazaspiro[4.5]dec-3-en-2-one ≥95%
3-Phenyl-1,4,8-triazaspiro[4.5]dec-3-en-2-one

Hazard: Irritant

Formula: C₁₃H₁₅N₃O
MDL Number: MFCD06751102
CAS Number: 887220-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR