Ethyl-3-[(4-methylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

115075-500MG 115075-1G
102898-946EA 693.85 USD
102898-946 102898-948
Ethyl-3-[(4-methylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate ≥97%
Ethyl-3-[(4-methylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₈N₄O₄S
MW: 302.35 g/mol
MDL Number: MFCD18194691
CAS Number: 1106908-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR