Ethyl-3-[(4-isopropylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

115074-500MG 115074-1G
102898-942EA 693.85 USD
102898-942 102898-944
Ethyl-3-[(4-isopropylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate ≥97%
Ethyl-3-[(4-isopropylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate
Formula: C₁₃H₂₂N₄O₄S
MDL Number: MFCD18194693
CAS Number: 1260986-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR