Ethyl-3-[(4-ethylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

115073-500MG 115073-1G
102898-938EA 693.85 USD
102898-938 102898-940
Ethyl-3-[(4-ethylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate ≥95%
Ethyl-3-[(4-ethylpiperazin-1-yl)sulfonyl]-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₀N₄O₄S
MW: 316.38 g/mol
MDL Number: MFCD18194692
CAS Number: 1240169-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR