6-Amino-1-(2-methoxyphenyl)pyrimidine-2,4(1H,3H)-dione ≥90%

Supplier: Matrix Scientific

115071-500MG 115071-1G
102895-354EA 693.85 USD
102895-354 102895-356
6-Amino-1-(2-methoxyphenyl)pyrimidine-2,4(1H,3H)-dione ≥90%
6-Amino-1-(2-methoxyphenyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₃
MDL Number: MFCD06486852
CAS Number: 180028-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR