2-(3-Fluorophenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥97%

Supplier: Matrix Scientific

115069-500MG 115069-1G
102895-346EA 693.85 USD
102895-346 102895-348
2-(3-Fluorophenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥97%
2-(3-Fluorophenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one

Hazard: Irritant

Formula: C₁₂H₈FN₃O
MDL Number: MFCD25951877
CAS Number: 1338651-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR