2-(4-Fluorophenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥95%

Supplier: Matrix Scientific

115067-500MG 115067-1G
102895-338EA 693.85 USD
102895-338 102895-340
2-(4-Fluorophenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥95%
2-(4-Fluorophenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one

Hazard: Irritant

Formula: C₁₂H₈FN₃O
MDL Number: MFCD11045716
CAS Number: 1338692-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR