8-[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl][1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥97%

Supplier: Matrix Scientific

115065-500MG 115065-1G
102895-330EA 693.85 USD
102895-330 102895-332
8-[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl][1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥97%
8-[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl][1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

Hazard: Irritant

Formula: C₁₄H₈ClN₅O₂
MW: 313.71 g/mol
MDL Number: MFCD14703518
CAS Number: 1239728-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR