1-(4-Bromo-3-methylphenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

115062-500MG 115062-1G
102895-324EA 693.85 USD
102895-324 102895-326
1-(4-Bromo-3-methylphenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid ≥97%
1-(4-Bromo-3-methylphenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀BrN₃O₂
MDL Number: MFCD11650614
CAS Number: 933242-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR