1-(2-Methoxyethyl)-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one ≥97%

Supplier: Matrix Scientific

115057-500MG 115057-1G
102891-280EA 693.85 USD
102891-280 102891-282
1-(2-Methoxyethyl)-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one ≥97%
1-(2-Methoxyethyl)-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD20528978
CAS Number: 1340857-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR