3-(3,4-Dimethylphenyl)-2-thioxo-2,3-dihydrothieno[3,2-d]pyrimidin-4(1H)-one ≥97%
Supplier: Matrix Scientific
115053-500MG
115053-1G
102895-306EA
693.85
USD
102895-306
102895-308
3-(3,4-Dimethylphenyl)-2-thioxo-2,3-dihydrothieno[3,2-d]pyrimidin-4(1H)-one ≥97%
3-(3,4-Dimethylphenyl)-2-thioxo-2,3-dihydrothieno[3,2-d]pyrimidin-4(1H)-one
Hazard: Irritant
Formula:
C₁₄H₁₂N₂OS₂ |
MDL Number:
MFCD03018748 CAS Number: 440327-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...