1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one ≥97%

Supplier: Matrix Scientific

115050-500MG 115050-1G
102895-294EA 693.85 USD
102895-294 102895-296
1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one ≥97%
1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one

Hazard: Irritant

Formula: C₁₅H₂₁N₃O
MW: 259.35 g/mol
MDL Number: MFCD20528963
CAS Number: 1351398-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR