2-(Methylthio)quinazolin-4(3H)-one ≥90%

Supplier: Matrix Scientific

115049-500MG 115049-1G
102895-290EA 693.85 USD
102895-290 102895-292
2-(Methylthio)quinazolin-4(3H)-one ≥90%
2-(Methylthio)quinazolin-4(3H)-one

Hazard: Irritant

Formula: C₉H₈N₂OS
MDL Number: MFCD00463044
CAS Number: 54855-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR