2-Azido-N-(2-ethyl-6-methylphenyl)acetamide ≥97%
Supplier: Matrix Scientific
115044-500MG
115044-1G
102895-270EA
693.85
USD
102895-270
102895-272
2-Azido-N-(2-ethyl-6-methylphenyl)acetamide ≥97%
2-Azido-N-(2-ethyl-6-methylphenyl)acetamide
Hazard: Irritant
Formula:
C₁₁H₁₄N₄O MW: 218.26 g/mol |
MDL Number:
MFCD14652806 CAS Number: 1249964-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...