2-Chloro-5-[3-(2-methoxyethyl)-1,2,4-oxadiazol-5-yl]pyridine ≥90%
Supplier: Matrix Scientific
115042-500MG
115042-1G
102895-262EA
693.85
USD
102895-262
102895-264
2-Chloro-5-[3-(2-methoxyethyl)-1,2,4-oxadiazol-5-yl]pyridine ≥90%
2-Chloro-5-[3-(2-methoxyethyl)-1,2,4-oxadiazol-5-yl]pyridine
Hazard: Irritant
Formula:
C₁₀H₁₀ClN₃O₂ MW: 239.66 g/mol |
MDL Number:
MFCD16875000 CAS Number: 1239723-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...