7-Methyl-6-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

115038-1G 115038-500MG
102895-248EA 564.62 USD
102895-248 102898-936
7-Methyl-6-nitro-1H-indazole ≥95%
7-Methyl-6-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Melting Pt: 221…225 °C
MDL Number: MFCD16620021
CAS Number: 208457-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR