6,7-Dimethoxy-4-(4-methylphenyl)naphthalene-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

115036-1G 115036-500MG
102898-930EA 843.08 USD
102898-930 102899-066
6,7-Dimethoxy-4-(4-methylphenyl)naphthalene-2-carboxylic acid ≥97%
6,7-Dimethoxy-4-(4-methylphenyl)naphthalene-2-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₈O₄
MW: 322.36 g/mol
Melting Pt: 250…251 °C
MDL Number: MFCD03212115
CAS Number: 109933-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR