Methyl-2-(4-chlorophenyl)-4-oxo-4-phenylbutanoate ≥97%

Supplier: Matrix Scientific

115035-500MG 115035-1G
102899-062EA 624.62 USD
102899-062 102899-064
Methyl-2-(4-chlorophenyl)-4-oxo-4-phenylbutanoate ≥97%
Methyl-2-(4-chlorophenyl)-4-oxo-4-phenylbutanoate

Hazard: Irritant

Formula: C₁₇H₁₅ClO₃
MW: 302.76 g/mol
MDL Number: MFCD28023506
CAS Number: 1383253-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR