6-(4-Nitrophenyl)imidazo[2,1-b][1,3]thiazole ≥95%

Supplier: Matrix Scientific

115030-500MG 115030-1G
102895-244EA 312.31 USD
102895-244 102895-246
6-(4-Nitrophenyl)imidazo[2,1-b][1,3]thiazole ≥95%
6-(4-Nitrophenyl)imidazo[2,1-b][1,3]thiazole

Hazard: Irritant

Formula: C₁₁H₇N₃O₂S
MDL Number: MFCD00113357
CAS Number: 7120-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR