Ethyl-N-[(5-methylpyridin-2-yl)carbamothioyl]carbamate ≥97%

Supplier: Matrix Scientific

115029-500MG 115029-1G
102895-240EA 312.31 USD
102895-240 102895-242
Ethyl-N-[(5-methylpyridin-2-yl)carbamothioyl]carbamate ≥97%
Ethyl-N-[(5-methylpyridin-2-yl)carbamothioyl]carbamate

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂S
MDL Number: MFCD18431230
CAS Number: 37890-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR