6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole ≥97%

Supplier: Matrix Scientific

115028-500MG 115028-1G
102895-236EA 290.77 USD
102895-236 102895-238
6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole ≥97%
6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₃S
MDL Number: MFCD06410799
CAS Number: 43036-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR