Ethyl-2-cyano-3-(3-cyanophenyl)prop-2-enoate ≥97%

Supplier: Matrix Scientific

115027-500MG 115027-1G
102895-232EA 204.62 USD
102895-232 102895-234
Ethyl-2-cyano-3-(3-cyanophenyl)prop-2-enoate ≥97%
Ethyl-2-cyano-3-(3-cyanophenyl)prop-2-enoate

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₂
MDL Number: MFCD03412730
CAS Number: 18153-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR