1-(4-Methanesulfonylphenyl)pentane-1,4-dione ≥97%

Supplier: Matrix Scientific

115024-500MG 115024-1G
102895-220EA 423.08 USD
102895-220 102895-222
1-(4-Methanesulfonylphenyl)pentane-1,4-dione ≥97%
1-(4-Methanesulfonylphenyl)pentane-1,4-dione

Hazard: Irritant

Formula: C₁₂H₁₄O₄S
MW: 254.31 g/mol
MDL Number: MFCD09030703
CAS Number: 189501-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR