2-Bromo-5-fluoroisonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-fluoro-4-pyridinecarboxylic acid, 2-Bromo-5-fluoropyridine-4-carboxylic acid

115277-1G 115277-5G
102894-626EA 267.69 USD
102894-626 102894-628
2-Bromo-5-fluoroisonicotinic acid ≥97%
2-Bromo-5-fluoroisonicotinic acid

Hazard: Irritant

Formula: C₆H₃BrFNO₂
MW: 219.9959 g/mol
Boiling Pt: 406°C
Flash Pt: 199°C
MDL Number: MFCD09261246
CAS Number: 885588-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR