2-Bromo-3-fluoroisonicotinic acid ≥97%

Supplier: Matrix Scientific

115268-1G 115268-5G
102894-510EA 60 USD
102894-510 102894-512
2-Bromo-3-fluoroisonicotinic acid ≥97%
2-Bromo-3-fluoroisonicotinic acid

Hazard: Irritant

Formula: C₆H₃BrFNO₂
Density: 1.903 g/cc
MDL Number: MFCD13185796
CAS Number: 1211530-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR