tert-Butyl-N-(2-cyanoethyl)carbamate ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Aminopropanenitrile, N-BOC protected

115019-1G 115019-10G
102895-210EA 253.85 USD
102895-210 102895-212
tert-Butyl-N-(2-cyanoethyl)carbamate ≥97%
tert-Butyl-N-(2-cyanoethyl)carbamate

Hazard: Irritant

Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
MDL Number: MFCD02180880
CAS Number: 53588-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR