3,4-Ethylenedioxythiophene ≥97%
Supplier: Matrix Scientific
Synonyms:
2,3-Dihydrothieno[3,4-b][1,4]dioxine, EDOT
115011-1G
115011-10G
102895-192EA
60
USD
102895-192
102895-194
3,4-Ethylenedioxythiophene ≥97%
3,4-Ethylenedioxythiophene
Hazard: Irritant
Formula:
C₆H₆O₂S MW: 142.17 g/mol |
MDL Number:
MFCD02093622 CAS Number: 126213-50-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...