5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile ≥97%

Supplier: Matrix Scientific

114960-1G 114960-10G
102895-094EA 253.85 USD
102895-094 102895-096
5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile ≥97%
5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile

Hazard: Irritant

Formula: C₈H₇ClN₂O
MDL Number: MFCD00052652
CAS Number: 23819-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR