Ethyl benzimidate hydrochloride ≥97%

Supplier: Matrix Scientific

114914-1G 114914-10G 114914-50G
102894-964EA 38.46 USD
102894-964 102894-966 102894-968
Ethyl benzimidate hydrochloride ≥97%
Ethyl benzimidate hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO
Melting Pt: 115…117 °C
MDL Number: MFCD00043246
CAS Number: 5333-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR