Diphenhydramine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Benzhydryloxy)-N,N-dimethylethanamine hydrochloride, Diphenhydramine HCl, 2-(Benzhydryloxy)-N,N-dimethylethylamine Hydrochloride, 2-Diphenylmethoxy-N,N-dimethylethylamine Hydrochloride

114865-10G 114865-50G
102894-842EA 67.69 USD
102894-842 102894-844
Diphenhydramine hydrochloride ≥97%
Diphenhydramine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₁NO
MW: 291.82 g/mol
MDL Number: MFCD00012479
CAS Number: 147-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR