3-Bromo-4-methylthiophene ≥97%

Supplier: Matrix Scientific

114768-1G 114768-10G
102898-902EA 156.92 USD
102898-902 102898-904
3-Bromo-4-methylthiophene ≥97%
3-Bromo-4-methylthiophene

Hazard: Irritant

Formula: C₅H₅BrS
MDL Number: MFCD00130084
CAS Number: 30318-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR