Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

114684-1G 114684-10G
102894-504EA 146.15 USD
102894-504 102894-506
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate ≥97%
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₃S
MW: 283.74 g/mol
MDL Number: MFCD00816787
CAS Number: 263016-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR