1-Methylpiperidine-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
114546-1G
114546-10G
102894-266EA
183.08
USD
102894-266
102894-268
1-Methylpiperidine-2-carboxylic acid ≥97%
1-Methylpiperidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₇H₁₃NO₂ Melting Pt: 216…221 °C |
MDL Number:
MFCD08060082 CAS Number: 7730-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...